Tlačiť
VWR®, Volumetric reagents for water determination by the Karl Fischer method
Dodávateľ: VWR Chemicals
85450.290EA 195.47 EUR
85450.290 85451.290 85452.290 85453.290 85454.290 85455.290 85456.290 85457.290 85458.290 85459.290 85461.290 85462.290 85463.290 85464.290 85465.290 85466.290 85468.290 85469.290 85453.320 85451.320 85452.320 85457.320 85463.320 85499.290
VWR®, Volumetric reagents for water determination by the Karl Fischer method
Karl Fischer titranty a činidla
These reagents are ideal for water determination including the analysis of aldehydes and ketones.
  • Fast and stable endpoints for accurate results time and time again
  • Low toxicity and pyridine-free
  • Provide consistent results