Tlačiť
Protein blocking agent, BLOT-QuickBlocker™
Dodávateľ: G Bioscienes
786-011
786-011EA 119.77 EUR
786-011
Protein blocking agent, BLOT-QuickBlocker™
Pufry Pufry pro elektroforézu Pufry blokovací
BLOT-QuickBlocker™ is a modified milk protein that is highly soluble and does not inhibit peroxidase detection. The modified milk protein has high blocking efficiency with a clear background.
  • Readily soluble and produces semi-clear solution
  • No inhibition to peroxidase
  • Fat free and produces clear background
  • Higher blocking efficiency
  • Blocking time 30 to 60 minutes