Tlačiť
Screw caps with silicone inserts, Top Works™ system, Nalgene®
Dodávateľ: Thermo Fisher Scientific
2135-3803 2135-5300 2135-5302 2135-5303 2135-8300 2135-8302 2135-8303
734-5043EA 405.37 EUR
734-5043 734-5044 734-5045 734-5046 734-5047 734-5048 734-5049
Screw caps with silicone inserts, Top Works™ system, Nalgene®
Uzávěry Uzávěry k láhvím
PP, white, with platinum cured silicone inserts.
  • Very good chemical resistance, heat resistant up to +135 °C, autoclavable
  • The Top Works™ system includes versions with solid, two or three port connections
  • Suitable for use in the aseptic transfer of liquids from most Nalgene® bottles and carboys
Platinum cured silicone tubing is fused through the stopper to form a single piece closure system.