Tlačiť
Avantor® Rigid Outer Support Containers
Dodávateľ: VWR Collection
734-3421EA 1746.04 EUR
734-3421 734-3422 734-3423 734-3424 734-3425 734-3426 734-3427 734-3428 734-3429 734-3430 734-3431 734-3432
Avantor® Rigid Outer Support Containers
Sáčky Sáčky skladovací
3D cubic bags can fit in existing support containers. These support containers are available in either High Density Polyethylene (HDPE) and Polypropelene (PP), or stainless steel material.
  • Standard 3D cubic bags are available in 100, 200, and 1000 L volumes with 3 top ports
  • Standard 3D cubic bags are available with or without a bottom port
  • Cleanroom and shipping variants are available in either material format
  • Plastic containers can collapse for easy storage
  • Stainless steel containers are ISTA certified for shipping
Avantor® 3D cubic bags are madeMore Product Information