Tlačiť
VWR®, Cell Culture Flasks, Non Treated or Treated
Dodávateľ: VWR Collection
734-2311EA 223.51 EUR
734-2311 734-2310 734-2312 734-2313 734-2314 734-2315 734-2316 734-2309 734-2600 734-2601 734-2783 734-2784 734-2785 734-2786 734-2787 734-2788 734-2789 734-2790 734-2791 734-2792 734-2804 734-2805 734-2806 734-2807 734-2808 734-2809 734-2810 734-2811 734-2812 734-2813 734-3252 734-3253 734-3265 734-3266 734-3419 734-3420
VWR®, Cell Culture Flasks, Non Treated or Treated
Baňky Baňky pro buněčné kultury
These cell and tissue culture flasks made of PS are perfect for cell growth.
  • Available with five different growth areas
  • Flask surface is flat and free from striations to maximise usable growth area
  • Two different HDPE cap styles can be used in both open and closed systems
  • Innovative angled neck design offers good pipette and cell scraper access
  • Triangular top with wide base provides stability
  • Easy stacking
  • Frosted area near the neck for easy marking/writing
  • Engraved graduations on multiple sides
  • 100% integrity tested
  • Sterilised by electron beam irradiation
More Product Information
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable Low Manufacturing Impact
Certifikát ochrany životního prostředí ISO 14001/Certificate No.: 04923E00243R0M