Tlačiť
Multi-layer cell culture flasks, Falcon® Multi-Flask
Dodávateľ: Corning
353144 353143
734-2457EA 201.08 EUR
734-2457 734-2456
Multi-layer cell culture flasks, Falcon® Multi-Flask
Baňky Baňky pro buněčné kultury
PS, tissue culture-treated, with membrane filter cap, non-pyrogenic.
  • Even distribution of the media across all layers, proven vacuum-gas tissue culture surface treatment and effective gas exchange all combine to provide an optimal cell culture environment
  • Cells can be easily accessed by pipette
  • Batch number printed on each individual flask for improved traceability
  • Manufactured in compliance with cGMP standards
Cell culture treated Falcon® Multi-Flasks are available with 3 or 5 layers and cell growth surfaces 525 or 875 cm². They provide greaterMore Product Information