Tlačiť
Centrifuge tubes, conical bottom, Falcon®
Dodávateľ: Corning
352070 352075 352076 352095 352096 352097 352098 352099
734-0448EA 227.12 EUR
734-0448 734-0449 734-0987 734-0450 734-0451 734-0452 734-0453 734-0454
Centrifuge tubes, conical bottom, Falcon®
Zkumavky Zkumavky centrifugační
Falcon conical tubes are easy-to-use, high quality tools to protect your precious samples. State-of-the-art design and manufacturing create tubes engineered to provide high strength, wide temperature stability, and to perform in your critical applications.
  • Meets bioanalytical grade requirements
  • Provide unsurpassed performance in critical research applications
  • Industry leading centrifugation rating of 16000×g for 50 ml (polypropylene)
  • Easy-to-read graduations
  • Consistent biological and physical properties
  • Ideal for long-term cryogenic storage of
More Product Information