Tlačiť
Poly-lysine culture dishes, Corning® BioCoat™
Dodávateľ: Corning
354468 354517 356468 356517 356518 354467 354469 354518 356467 356469
734-0149EA 132.99 EUR
734-0149 734-0177 734-0284 734-0301 734-0302 734-0148 734-0150 734-0178 734-0417 734-0285
Poly-lysine culture dishes, Corning® BioCoat™
Misky kultivační Misky pro tkáňové kultury
PS, coated with poly-lysine.
Poly-D-lysine (PDL) and Poly-L-lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. Poly-lysine surface treatments support applications including attachment and spreading of a variety of cell lines; cell differentiation and neurite outgrowth; attachment of transfected cell lines; and survival of primary neurons in culture.