Tlačiť
Poly-D-Lysine flasks, Corning® BioCoat®
Dodávateľ: Corning
356538 356539 354536 354537 354538 354539 356536 356537
734-0312EA 813.94 EUR
734-0312 734-0419 734-0191 734-0192 734-0193 734-0194 734-0311 734-0418
Poly-D-Lysine flasks, Corning® BioCoat®
Baňky Baňky pro buněčné kultury
Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.
  • Attachment and spreading of a variety of cell types
  • Cell
More Product Information