Tlačiť
Filter tips, Thermo Scientific™ ClipTip™
Dodávateľ: Thermo Fisher Scientific
94420213 94420218 94420253 94420258 94420313 94420318 94420513 94420518 94420713 94420718 94420043 94420053 94420103 94420153 94420813 94420818 94420613 94420063 94420073
732-1618EA 69.9 EUR
732-1618 732-1619 732-1620 732-1621 732-1622 732-1623 732-1624 732-1625 732-1626 732-1627 732-5053 732-5050 732-5051 732-5052 732-5054 732-5055 613-6197 732-3664 613-6196
Filter tips, Thermo Scientific™ ClipTip™
Špičky pipetovací
ClipTip filtered pipette tips provide security with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force.
  • Pipette tips feature 'clips' that lock tips in place and forms a complete seal
  • Every ClipTip pipette tip utilises low retention technology to enable maximium sample recovery
  • Certified free of RNase, DNase, DNA, ATP and endotoxin contamination
  • Environmentally friendly reload insert packaging uses less plastic
  • Each tip insert has ink-jetted lot and volume information;
More Product Information
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable Low Manufacturing Impact
Certifikát ochrany životního prostředí ISO 14001 (Certificate Registration No. EMS 623400)