Tlačiť
VWR®, Low Temperature PCR Rack, 96 Well
Dodávateľ: VWR Collection
732-2975EA 77.71 EUR
732-2975 732-3319 732-3320 732-3321
VWR®, Low Temperature PCR Rack, 96 Well
Stojany Tube Racks
This PCR storage rack offers a sleek and modern design that includes a unique lid that the rack slides out from, and then rotates 180°, to become a convenient stand that the rack then sits on. Ergonomic, textured finger grips provide a secure hold, reducing the chance of dropping the PCR rack, making handling and transportation easy, especially when using gloves.
  • Holds 0,2 ml tubes, individual or strip
  • 96-well (8×12 configuration)
  • For freezer storage down to –80 °C
  • Detachable lid
The lid has a side opening that enables faster cooling of samples toMore Product Information
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable