Tlačiť
PCR plates, 384-well, Armadillo™
Dodávateľ: Thermo Fisher Scientific
AB-3384 BC-2384 AB-3384/Y AB-3384/R AB-3384/O AB-2384 AB-3384/B BC-2384/O AB-2384/Y AB-2384/R AB-2384/O AB-2384/G AB-2384/B AB-3384/G BC-3384/G BC-3384/Y BC-2384/B BC-3384/O BC-2384/G BC-3384/B BC-3384 BC-2384/Y BC-2384/R BC-3384/R
732-1579EA 455.84 EUR
732-1579 732-1583 732-2808 732-2807 732-1582 732-1575 732-1580 732-1586 732-2792 732-2791 732-1578 732-1577 732-1576 732-1581 732-1589 732-2822 732-1584 732-1590 732-1585 732-1588 732-1587 732-2800 732-2799 732-2821
PCR plates, 384-well, Armadillo™
Mikrodestičky Destičky pro PCR
Armadillo™ 384-well PCR plates combine the rigidity of a polycarbonate frame with thin walled polypropylene wells to provide superior thermal cycling performance under all conditions, without warping. All formats are available with clear or white wells, ensuring the highest level of sensitivity for all PCR and qPCR reactions.
  • Optically clear skirt aids visualisation of liquids and aids stacking in automated handling systems
  • Raised rim design gives improved well surface area and improves seal integrity in automated heat sealing
  • SBS footprint ensures