Tlačiť
Barrier tips, extended, ART™
Dodávateľ: Thermo Fisher Scientific
2140-HR 2079-HR 2080-HR 2160P-HR
732-1140EA 72.14 EUR
732-1140 732-2398 732-2400 732-2406
Barrier tips, extended, ART™
Špičky pipetovací
These barrier tips are designed to fit virtually any pipette and application. The self-sealing barrier provides 100% protection against aerosols and liquid contamination.
  • Transparent, sterile
  • Free of RNase, DNase, DNA, ATP, Bioburden, Endotoxin, Pyrogen and PCR Inhibitors
  • Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
  • Rack cover can easily be realigned from a traditional lift-off to a hinged rack for one-handed access to tips
  • Front latch closes and locks, ensuring safe and secure storage
Limited Time Sale
Select items below are currently on sale
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable Low Manufacturing Impact
Certifikát ochrany životního prostředí IRECS; ISO 14001 (Certificate Registration No. EMS 752603)