Tlačiť
VWR® 96-Well Deep-Well Plates with Automation Notches
Dodávateľ: VWR Collection
732-0585EA 65.29 EUR
732-0585
VWR® 96-Well Deep-Well Plates with Automation Notches
Mikrodestičky Destičky s hlubokými jamkami
Heavy polypropylene deep well plates feature square wells, molded alphanumeric labeling, and smooth interior surfaces to ensure that all the samples can be pipetted.
  • Alphanumeric indexing
  • Square well with V-bottom design
  • Autoclavable
  • Freezable to –80 °C (–112 °F)
VWR® deep-well plates feature printed alphanumeric labeling and smooth interior surfaces to ensure easy pipetting. Plates and mats are free of RNase, DNase and human DNA (lot-by-lot certificates available).
Applications include HTS, combinatorial chemistry, library applications (compound