Tlačiť
Ultra clear cap strips for qPCR
Dodávateľ: Thermo Fisher Scientific
AB-0866
732-0081EA 72.9 EUR
732-0081
Ultra clear cap strips for qPCR
Zkumavky Zkumavky pro PCR
PP flat 8-cap strips designed for qPCR and other fluorescence applications.
  • Compatible with all Thermo Scientific™ 96-well PCR plates, as well as standard and low profile tube strips
  • Specially formulated ultra clear polypropylene enhances signal detection by minimising autoflourescence and maximising transmission
  • Highly polished flat surface provides an optically clear window to deliver maximum and consistent signal transmission, critical for accurate qPCR results
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable