Tlačiť
PCR plates, 96-well, Armadillo™
Dodávateľ: Thermo Fisher Scientific
AB-3396/Y AB-3396/R AB-3396/O AB-3396/G AB-3396/B AB-3396 AB-2396/Y AB-2396/R AB-2396/O AB-2396/G AB-2396/B AB-2396 BC-3396/Y BC-3396/R BC-3396/O BC-3396/G BC-3396/B BC-3396 BC-2396/Y BC-2396/R BC-2396/O BC-2396/G BC-2396/B BC-2396 BC-2496 AB-2496 AB-2496/B AB-2496/G AB-2496/O AB-2496/R AB-2496/Y AB-3496 AB-3496/B AB-3496/G AB-3496/O AB-3496/R AB-3496/Y BC-2496/B BC-2496/O BC-2496/R BC-2496/Y BC-3496 BC-3496/B BC-3496/O BC-3496/R AB-2596 AB-2596/B AB-2596/G AB-2596/O AB-2596/R AB-2596/Y AB-3596 AB-3596/B AB-3596/G AB-3596/O AB-3596/R AB-3596/Y BC-2596 BC-2596/G BC-2596/O BC-2596/R BC-2596/Y BC-3596 BC-3596/G BC-3596/O BC-3596/R BC-3596/Y BC-2596/B
731-0377EA 171.44 EUR
731-0377 731-0376 731-0375 731-0374 731-0373 731-0364 731-0372 731-0371 731-0370 731-0369 731-0368 731-0363 731-0389 731-0388 731-0387 731-0386 731-0385 731-0384 731-0383 731-0382 731-0381 731-0380 731-0379 731-0378 732-2801 732-2793 732-2794 732-2795 732-2796 732-2797 732-2798 732-2809 732-2810 732-2811 732-2812 732-2813 732-2814 732-2802 732-2804 732-2805 732-2806 732-2823 732-2824 732-2826 732-2827 731-0644 731-0655 731-0645 731-0646 731-0647 731-0648 732-2815 732-2816 732-2817 732-2818 732-2819 732-2820 731-0649 731-0650 731-0651 731-0653 731-0654 732-2829 732-2831 732-2832 732-2833 732-2834 731-0652
PCR plates, 96-well, Armadillo™
Mikrodestičky Destičky pro PCR
Rigid PC frame, thin-walled PP wells. Ultra-rigid 96-well PCR plates with rounded V-bottom wells designed for robotic applications.
  • Optically clear skirt aids visualisation of liquids and improves stacking capabilities in automated handling systems
  • Raised rim design improves seal integrity in automated heat sealing
  • With a rigid polycarbonate skirt, the ANSI/SBS footprint ensures complete compatibility with robotic systems for high throughput applications
  • Thermostable polycarbonate frame design allows warp-resistant thermal cycling and enhanced mechanical
More Product Information
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable