Tlačiť
Deep well plates, 96-well, Bioblock™
Dodávateľ: SIMPORT SCIENTIFIC
T110-6
730-1091EA 229.52 EUR
730-1091
Deep well plates, 96-well, Bioblock™
Mikrodestičky Destičky s hlubokými jamkami
PS. The 1,2 ml capacity round bottom deep well plate (1 ml when capped) can withstand centrifugation up to 3000×g. It is not DMSO resistant.
Informace o dodávce: Packed in bags of 4 plates.
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable