Tlačiť
UV to white light converter plates
Dodávateľ: UVP ULTRA VIOLET PRODUCTS
38-0191-01 38-0191-02 38-0191-04 38-0191-11
730-1017EA 476.27 EUR
730-1017 730-1018 UVPA38-0191-04 UVPA38-0191-11
UV to white light converter plates
Transiluminátory Transiluminátory s UV/bílým světlem
The specially designed converter plate converts UV radiation from the transilluminator using a white light diffuser to enable Coomassie® Blue or silver stained protein gels to be viewed.
  • Handles are situated on opposite sides of the converter plate for easy handling