Tlačiť
Infusion filter, Sterifix® Paed
Dodávateľ: BRAUN
4099354
720-2580EA 907.27 EUR
720-2580
Infusion filter, Sterifix® Paed
Infuze
Sterifix Paed is specially designed for the bacteria and particle retention.
  • Reliable air venting regardless of position
  • Flow rate (aqua dest.) over 10 ml per minute
  • Luer Lock fitting