Tlačiť
Dissection boards, DissecTable™
Dodávateľ: SIMPORT SCIENTIFIC
M618 M620 M625
720-1499EA 203.48 EUR
720-1499 720-1685 720-1686
Dissection boards, DissecTable™
Desky krájecí
HDPE / PS. These dissection boards feature a large cutting area on one side. They include dimensional scales in inches and centimeters, a grid and dimensional circles of different sizes. The other side offers two cutting boards half the size with the same dimensional features printed on each one of them.
  • Drain groove is carved all around the edge of the boards in order to contain fluids
  • All corners have rubber feet giving more stability to the working surface
  • Bases are stackable and will not move sideways during dissecting work
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable