Tlačiť
Food safe kits for detecting animal species in meat products
Dodávateľ: BIOPREMIER
FS-0006 FS-0007 FS-0008 FS-0009 FS-0010 FS-0011
710-1490EA 704.59 EUR
710-1490 710-1491 710-1492 710-1493 710-1494 710-1495
Food safe kits for detecting animal species in meat products
Mikrobiologické testovací systémy PCR Testy
Food safe kits provide a simple, reliable, and rapid procedure for detecting the presence of an animal species in meat and meat products. The assay is based on 5’ nuclease Real-Time PCR reactions to amplify a unique genomic sequence in the target organism. According to European Commission Directive 2002/86/EC, food ingredients have to be declared. In the case of meat and meat products, the animal species has to be disclosed on the label. Moreover, the species autenticity can be highly relevant to consumers for economic, medical, cultural and religious reasons. Fraudulent substitution of cheaper meats in place of more expensive species, the inclusion of meat in non meat (vegetarian) products and the presence of allergens in food products are clear examples for the importance of this issue (Ballin, 2010; Dooley et al., 2004). Several methods are based on identification of proteins by means of electrophoretic and/or immunological methods. However, these methods are not reliable for application in highly processed and heated products due to the protein deterioration. DNA is more stable than proteins during processing and although it can be fragmented by several processes, modern DNA methodologies like PCR-based techniques still allow the identification of DNA from the different species present in a sample (Lockley & Bardsley, 2000). Real-Time PCR techniques are especially suitable for these products because small fragments of DNA can still be amplified and identified with high sensitivity and specificity (Dooley et al., 2004).
  • Ready to use and complete: All reagents for Real-Time PCR are included
  • Compatible with instruments working in FAM and ROX channels
  • Results obtained in about 2 hours after DNA extraction
  • Six kits for six different animal species
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable