Tlačiť
Contact plate and Petri dish reader, SP Bel-Art
Dodávateľ: Bel-Art Products, a Part of SP
F378610000
710-0079EA 396.15 EUR
710-0079
Contact plate and Petri dish reader, SP Bel-Art
Misky kultivační Misky Petriho
Made of polypropylene with a glass-clear acrylic deck for placement of plates and Petri dishes, this reader allows safe examination of the bottom of plates without risking spills on the face and eyes.
  • Ideal for examining cell monolayer, cell clones and for counting viral plaques or bacterial colonies
  • Easy and safe reading of hemagglutination, hemagglutination inhibition, hemolysis and agglutination tests
  • Clear deck for resting plates
  • Large, adjustable mirror is perfect for viewing plate underside
  • Mirror magnification is 3X actual plate size