Tlačiť
Kjeldahl distillation system, VAPODEST® 200
Dodávateľ: GERHARDT
12-0520
705-0857EA 0 EUR
705-0857
Kjeldahl distillation system, VAPODEST® 200
Destilační systémy Kjeldahlův destilační systém
VAPODEST® 200 is the ideal instrument for distillation of volatiles, SO₂, alcohol and phenol.
  • Intelligent supervision of all vital functions
  • Programmable, automatic addition of NaOH
  • Programmable distillation and reaction time
  • Efficient with programmable steam power 10 to 100%
  • 7" TFT fully coloured display and easy menu navigation
VAPODEST® 200 complies with the data management requirements of accredited laboratories (ISO 17025, GLP).