Tlačiť
COD test kits and consummables for AQUAfast COD colorimeters
Dodávateľ: Thermo Orion
CODHP0 CODH00 CODL00
662-1543EA 87.72 EUR
662-1543 705-0757 705-0758
COD test kits and consummables for AQUAfast COD colorimeters
Kity pro environmentální testy
Orion™ AQUAfast™ COD reagents in digestion tubes are designed for use with Orion™ AQUAfast™ colorimeters and Orion™ AquaMate Vis and UV-Vis spectrophotometers. Just add the sample to the COD digestion tube, digest sample in thermoreactor and measure.
  • Reagents are fully prepared and offer convenience and added safety for user
  • Reagents comply with EPA guidelnes
Digestion tubes contain the precise amount of reagent, are made of special optical glass, and fit the Orion™ AQUAfast™ COD165 thermoreactor.