Tlačiť
Peqlab PerfectBlue Midi ExW, Wide Format Gel System Midi ExW
Dodávateľ: VWR Peqlab
700-0531EA 0 EUR
700-0531
Peqlab PerfectBlue Midi ExW, Wide Format Gel System Midi ExW
Systémy elektroforetické Systémy pro horizontální gelovou elektroforézu
With its extra wide design, the Midi ExW is ideal when there is a need to run lots of samples (up to 200) just a short distance, for example when genotyping. This format, along with its compact footprint, offers rapid run times without compromising on power. The result is a system for high throughput sample processing generating crisp clear band data. As with all PerfectBlue™ gel systems, gels can be poured directly in the trays at temperatures up to 60 °C. With the Midi ExW, the trays can be walled with effective gasketed 'end gates' which are supplied in the system. Additionally, gel sizeMore Product Information