Tlačiť
VWR® shiroGEL vertical 20, Vertical Electrophoresis Systems
Dodávateľ: VWR Collection
700-0353EA 2398.16 EUR
700-0353 700-0354
VWR® shiroGEL vertical 20, Vertical Electrophoresis Systems
Systémy elektroforetické Systémy pro vertikální gelovou elektroforézu
The shiroGEL vertical 20 offers application flexibility and is distinguished by its ease of gel casting and set-up, requiring only four screws to secure up to four 20×20 cm gels. This vertical screw clamp technology distributes pressure evenly along the height of gel rather than in the centre to eliminate plate bowing and gel compression, while maintaining a leakproof seal during casting. A built-in inner buffer chamber within the PAGE insert allows set up to be completed without inclusion of an upper buffer chamber. These systems are ideal for 1D and 2D SDS-PAGE, native, preparative,More Product Information