Tlačiť
Orion Star™ A329 Multi-Parameter Meter (pH/ISE/Conductivity/DO), Handheld
Dodávateľ: THERMO ORION
STARA3290 STARA3295
665-0281EA 2491.49 EUR
665-0281 665-0282
Orion Star™ A329 Multi-Parameter Meter (pH/ISE/Conductivity/DO), Handheld
Meřiče elektrochemické Meřiče pH a multimetry
This rugged, waterproof portable meter is designed for a wide range of pH, ion concentration, conductivity, dissolved oxygen and temperature testing and field applications. It can be used in the most demanding locations thanks to its IP67-rated housing. The meter offers three measuring channels allowing users to measure pH, conductivity and DO simultaneously or view each channel separately.
  • Up to five point pH calibrations with calibration editing option that allows individual points to be fixed without a full recalibration
  • Automatically compensate
More Product Information