Tlačiť
Probe maintenance kits for dissolved oxygen meters, Orion™
Dodávateľ: THERMO ORION
O80113 080513
662-0692EA 151.01 EUR
662-0692 664-0499
Probe maintenance kits for dissolved oxygen meters, Orion™
Elektrody Sondy pro stanovení kyslíku
These probe maintenance kits help ensure optimal performance of Orion™ polarographic dissolved oxygen probes.
Informace o dodávce: Each probe maintenance kit includes two membrane caps, one electrolyte solution, and a polishing disk.