Tlačiť
Orion Star™ A323 Dissolved Oxygen Meter, Handheld
Dodávateľ: THERMO ORION
STARA3230 STARA3235
664-0149EA 1301.02 EUR
664-0149 664-0150
Orion Star™ A323 Dissolved Oxygen Meter, Handheld
Meřiče elektrochemické Měřiče rozpušteného kyslíku
This portable DO/RDO meter is designed for a wide range of dissolved oxygen and temperature testing and field applications. It can be used with a RDO optical or a polarographic DO probe. The meter is suitable for use in the most demanding locations thanks to its IP67-rated housing.
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Measurement stability indicator and selectable read modes – auto-read holds a stable reading, continuous shows changing readings with hold option and timed logs data at