Tlačiť
TDS meter, handheld, Eutech TDS 6+
Dodávateľ: EUTECH
ECTDS603PLUS ECTDS603PLUSK
663-0354EA 586.82 EUR
663-0354 663-0355
TDS meter, handheld, Eutech TDS 6+
Meřiče elektrochemické Konduktometry
This portable economic meter with large LCD display is ideal for quick, accurate TDS checks in laboratories, field and schools.
  • Option of quick, easy automatic calibration or customised, near-to-sample manual calibration
  • Auto-ranging for conductivity measurements for fast response and best resolution over wide measurement range
  • HOLD function and auto-power off
  • Self-diagnostic with message codes for easy troubleshooting
  • Electrode with built-in ATC designed for minimal air bubble entrapment during measurement
  • Built with shock-resistant ABS plastic and
More Product Information