Tlačiť
Reference electrode, Orion™ 800500U ROSS Ultra™
Dodávateľ: THERMO ORION
800500U
662-1583EA 312.04 EUR
662-1583
Reference electrode, Orion™ 800500U ROSS Ultra™
Elektrody Elektrody pH
Half-cell reference electrode with glass body and pin-tip connector. Compatible with 8101BNWP, 8101SC ROSS pH half-cell and 8411BN ROSS sodium half-cell electrodes.
  • ROSS electrodes avoid the long term drift that other electrodes exhibit (ROSS electrodes drift by less than 0,002 pH per day), so recalibration is minimised
  • No sample contamination: ROSS electrodes do not contain any silver or mercury to react with the sample or to clog the electrode junction
  • Can be used in samples such as biological media, foodstuffs and pharmaceuticals where trace amount of