Tlačiť
Iontově selektivní elektrody, sodíkové, Orion™ ROSS™ Sure-Flow™
Dodávateľ: THERMO ORION
8411BN 8611BNWP
662-0747EA 613.66 EUR
662-0747 662-1523
Iontově selektivní elektrody, sodíkové, Orion™ ROSS™ Sure-Flow™
Elektrody Elektrody ISE
These electrodes are designed for measuring sodium in aqueous solutions quickly, accurately and economically.
  • Orion™ ROSS™ electrode provides faster response, greater stability and accuracy than conventional sodium electrodes
  • Combination electrode 8611BNWP does not require a separate reference electrode, making it convenient to use with small sample volumes
  • Built-in Sure-Flow reference junction prevents electrode clogging and provides fast and stabile readings
Objednávacie informácie: Tyto indikační elektrody vyžadují zvláštní referenční elektrodu,More Product Information