Tlačiť

Rat IgG2a Isotype Control [clone: 2A3] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)

Dodávateľ: TONBO BIOSCIENCE
65-4321-U100
65-4321-U100EA 124.17 EUR
65-4321-U100
Rat IgG2a Isotype Control [clone: 2A3] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Protilátky
The 2A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2a antibodies.

Type: Control
Antigen: IgG2a (Rat) (rat IgG2a)
Clonality: monoclonal
Clone: 2A3
Conjugation: Peridinin Chlorophyll/Cy5.5®
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR