Tlačiť
Marco Cells for Absorption Measurements with PTFE Lids, Hellma Analytics
Dodávateľ: HELLMA
6030-UV-10-531 6030-10-10 6030-20-10 6030-40-10 6030-50-10 AS2C-10K0-3M50 AS2C-40K0-14M0 AS5C-10K0-3M50
634-9005EA 103.75 EUR
634-9005 634-9001 634-9002 634-9003 634-9004 HELLAS2C-10K0-3M50 HELLAS2C-40K0-14M0 HELLAS5C-10K0-3M50
Marco Cells for Absorption Measurements with PTFE Lids, Hellma Analytics
Kyvety Kyvety spektrofotometrické
Hellma® macro cells for absorption measurements in the Vis range, 10 mm path length, with lid.
  • Cell type 6030-OG, from optical glass
  • Wavelength range from 260 to 2500 nm
  • 10 to 50 mm path length
  • With PTFE lid
  • Volume from 3,500 - 17,500 μl
Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers. Highest precision: Hellma® precision cells are characterised by the highest precision in path length accuracy and parallelism. This guarantees continuously reproducible measurement results.