Tlačiť
Spectrophotometric colorimeters, PFX995 series, Lovibond®
Dodávateľ: TINTOMETER
1379951 1379951H 1379952 1379952H
633-0238EA 21350.29 EUR
633-0238 633-0239 633-0240 633-0241
Spectrophotometric colorimeters, PFX995 series, Lovibond®
Kolorimetry Automatické kolorimetry pro měření transmitance
The PFX995 spectrophotometers are top of the range, versatile instruments that offer comprehensive colour data requirements for either the Petrochemicals industry or the Food and Chemicals industries for edible oils, fats, chemicals etc. The PFX995 and PFX995/P provide objective, unbiased colour data according to a comprehensive range of established industry scales, spectral data and CIE values. They are ideal for companies that process a broad selection of product types with varied colour specifications, particularly in central test facilities or in independent testingMore Product Information