Tlačiť
Slide boxes, SlideFile™
Dodávateľ: SIMPORT SCIENTIFIC
M700-100B M700-100G M700-100P M700-100W M700-100Y
631-1900EA 185.06 EUR
631-1900 631-1901 631-1902 631-1903 631-1904
Slide boxes, SlideFile™
Boxy na mikroskopická sklíčka
This impact resistant storage system can hold up to 400 slides per unit, for space efficient storage. All slides are stored upright for easier manipulation.
  • Removable tray inside the storage box for 100 individual numbered slots
  • Transparent hinged cover makes it easy to see the contents of the box at a glance
  • Unique feature with this system to read barcodes without having to remove the slides from the box
W×D×H: 82×245×86 mm
Objednávacie informácie: Each system includes a slide box and a removable tray.
Informace o dodávce: Index cards numberedMore Product Information