Tlačiť
Semi-automated rotary microtome, CUT 5062
Dodávateľ: Slee
10070001
631-0542EA 14972.16 EUR
631-0542
Semi-automated rotary microtome, CUT 5062
Mikrotomy Mikrotomy
The CUT 5062 is a semi-automated rotary microtome designed for paraffin sections.
  • Motorised trimming up to 750 μm and free programmable
  • Motorised advance/return of specimen
  • Automatic advance in two speeds
  • Variable retraction and section counter
  • Electric automatic stop function
Used in medical research, routine diagnostics or industrial applications, the modular design allows custom-made assemblies from a huge range of accessories.