Tlačiť
Cellcyte Gen 2 Live Cell Imaging and Analysis
Dodávateľ: DISCOVER ECHO
ECCG2230 ECCG2CU2
630-3561EA 69860.2 EUR
630-3561 630-3558
Cellcyte Gen 2 Live Cell Imaging and Analysis
Mikroskopy
Live cell imaging and analysis in real-time from within the incubator.
  • Enhanced phase contrast: High resolution and contrast rich transmission imaging mode
  • Fluorescence microscopy: Three channel (DAPI, GFP, mCherry) or four channel (DAPI, GFP, RFP, CY5) available
  • Open fanless design: easier accessibility and maintenance; uses convection to heat and circulate warm air, eliminating vibration and the formation of hot pockets
  • Acquisition software: Automated acquisition and user friendly image and data visualisation software
  • Analysis software: Integrated real-
More Product Information