Tlačiť
VWR® Traceable®, Temperature Data Loggers with TraceableGO™ Wireless Capability and Calibration
Dodávateľ: VWR Collection
620-2734EA 158.22 EUR
620-2734 620-2735 620-2736
VWR® Traceable®, Temperature Data Loggers with TraceableGO™ Wireless Capability and Calibration
Záznamníky dat (Dataloggery)
Easily access collected data between destinations with Bluetooth® data loggers.
  • Connect to your data via smartphone or tablet—any Bluetooth-enabled device—using a downloadable Bluetooth App
  • View and download data with free TraceableGO App
  • Data loggers store up to 90 alarm events
  • Readings and set-up parameters easily viewable on large 2" by 1" LCD
  • Get accurate, reliable measurements with included NIST traceable calibration
You have the option to subscribe to cloud-based TraceableLIVE® service to upload data to cloud—ideal for tracking data fromMore Product Information