Tlačiť
Flow indicator paddle wheel
Dodávateľ: SEMADENI
3228
618-1130EA 59.68 EUR
618-1130
Flow indicator paddle wheel
Průtokoměry
Flow indicator with propeller, Styrolacrylnitrile (SAN) transparent.
  • Corrosion resistant design without metal parts
  • Operating temperature up to 30 °C
  • Mounting length 90 mm
Suitable for gas and water flow control.
Pozor: Not suitable for use with solvents, acids or alkalis.