Tlačiť
Specific gravity cups, calibrated
Dodávateľ: Industrial Physics
VF2099 VF2100 VF2097 VF2098
614-0715EA 458.24 EUR
614-0715 SIMEVF2100 SIMEVF2097 SIMEVF2098
Specific gravity cups, calibrated
Pyknometry
Specific gravity cups for determining the specific gravity or density (or weight per gallon wpg) of coatings, pastes or similar liquids.
  • Density is defined as weight per unit volume at a specified temperature
  • Available in anodised aluminium or stainless steel
Certifikácie: According to ISO 2811, DIN 53 217 and ASTM D 1475.
Informace o dodávce: Supplied with a calibration certificate.