Tlačiť
Bottle-top dispenser, Calibrex™
Dodávateľ: Socorex Isba
520.002 520.005 520.010
613-4855EA 416.98 EUR
613-4855 613-4856 613-4857
Bottle-top dispenser, Calibrex™
Dávkovače na lahve
With fix or adjustable volume.
  • PFA coated plungerwith broad chemical resistance
  • QR coded chemical compatibility
  • Integrated calibration key
  • Quick disassembling, no tool required
  • Autoclavable at 121 °C fully assembled
This model is ranging 1 to 10 ml. The Calibrex™ combine a clever simple construction and a universal chemical compatibility. Appreciated for its smooth, effort-less activation and sturdiness, the base thread fits 32 mm bottle neck diameter.
Informace o dodávce: Supplied with three PP adapters for 28, 40 and 45 mm bottle neck diameters -More Product Information