Tlačiť
Combitips® advanced PCR Clean - Positive Displacement Pipette Tips
Dodávateľ: EPPENDORF
0030089.839 0030089.820 0030089.812 0030089.804 0030089.790 0030089.782 0030089.766 0030089.847 0030089.774
613-2083EA 186.26 EUR
613-2083 613-2082 613-2081 613-2080 613-2079 613-2078 613-2076 613-2084 613-2077
Combitips® advanced PCR Clean - Positive Displacement Pipette Tips
Špičky pipetovací
Eppendorf Combitips® advanced in 'PCR clean' grade offer superior positive displacement pipetting, ensuring precise handling of sensitive liquids in PCR and molecular biology, with contamination-free, ergonomic design for reliable lab results.
  • Combitips® advanced tips are the dedicated consumable for all Repeater®/Multipette® multi-dispenser pipettes from Eppendorf
  • 9 volume sizes (0,1 to 50 ml) offer a maximum range of dispensing volumes with increments as low as 100 nl
  • Certified to be free from human DNA, DNase, RNase, and PCR inhibitors that could
More Product Information