Tlačiť
VWR® Bulb Pipettes, One Mark, Class AS, with Single Certificate
Dodávateľ: VWR Collection
612-6712EA 36.37 EUR
612-6712 612-6713 612-6714 612-6715 612-6716 612-6717 612-6718 612-6719 612-6720 612-6721
VWR® Bulb Pipettes, One Mark, Class AS, with Single Certificate
Pipety Pipety nedělené
Borosilicate 9.1 glass. Single certified class AS bulb pipettes with one mark, imprinted serial number and individual work certificate.
  • Calibrated to deliver (TD, Ex)
  • Inscriptions and ring mark in high contrasting amber
  • Imprinted serial number, single certificate included
Made from true bore glass tubing to guarantee exact measurement at each measuring point. Colour coded and conformity certified with DE-M mark.
Certifikácie: DIN EN ISO 648