Tlačiť
VWR®, Časovač digitální, čtyřkanálový
Dodávateľ: VWR Collection
609-0194EA 75.31 EUR
609-0194 609-0226
VWR®, Časovač digitální, čtyřkanálový
Časovace/Stopky Časovače digitální
These digital timers allow the setting of four separate channels simultaneously in any combination of countdown or count-up (stopwatch).
  • Time-out mode stops the timer for off periods in both up and down timing
  • 12/24 hour 'time of day' clock
  • Large clear one-line 19 mm display allows viewing from across the lab
  • Capacity of 99 H, 59 min, 59 s in steps of 1 s (±0,01%)
  • Extra loud alarm for 1 min, or can be silenced manually
  • Unique memory displays previously programmed countdown time
When zero is reached, the timer starts counting up to show time elapsedMore Product Information