Tlačiť
b.safe Funnels 180
Dodávateľ: Bohlender
R800-02 R800-05 R800-10 R801-30 R810-05 R811-30 R820-05 R821-30 R830-05 R830-10 R831-30 R840-05 R840-10 R841-30 R850-05 R851-30 R860-05 R861-30 R870-05 R871-30 R880-05 R880-10 R890-05 R890-10
590-0358EA 143 EUR
590-0358 590-0359 590-0360 590-0362 590-0364 590-0366 590-0368 590-0371 590-0373 590-0374 590-0377 590-0379 590-0380 590-0387 590-0390 590-0393 590-0395 590-0397 590-0399 590-0400 590-0401 590-0402 590-0405 590-0406
b.safe Funnels 180
Nálevky Nálevky bezpečnostní
Funnel made of PE / static dissipative PE-EX for liquid waste disposal. The funnel can be safely mounted on the waste container with its freely turnable cap that is safely fixed with the filling neck.
  • Easy and safe handling: you do not need to open a lid before pouring
  • Removable sieve restrains, e.g. magnetic stirring bars or other solids
  • Splash guard for a smooth flow without splashing
  • Ball valve protects from harmful vapours; it floats up during filling and opens the drain
  • Free turnable cap for easy assembly on collecting container
  • Neck with plastic nibs
More Product Information