Tlačiť
Gas purifiers, GateKeeper® GPU 200 4R
Dodávateľ: ENTEGRIS
GPUS200FIX04R0 GPUS200FHX04R0 GPUS200FOP04R0
568-0913EA 0 EUR
568-0913 568-0912 568-0914
Gas purifiers, GateKeeper® GPU 200 4R
Filtry Filtry na plyny
GateKeeper® GPU gas purifiers were designed to improve purity consistency, lifetime and final purity. Leveraging its expertise in material science, Entegris has developed a new generation of purification products that deliver unsurpassed value and performance.
  • Purity control to ppt levels
  • Low pressure drop across the purifier
  • Regenerable
  • Long lifetime
  • Use ambient temperature purification – no heat or power required to operate
  • Insensitive to orientation (horizontal/vertical)
The model 200 is designed for peak flow rates up to 20 slpm.
Certifikácie: More Product Information