Tlačiť
Industrial label printers with Brady workstation LAB suite, BradyPrinter i7100
Dodávateľ: BRADY
306883 306886 306889 306880
568-0897EA 3459.24 EUR
568-0897 568-0899 568-0901 568-0895
Industrial label printers with Brady workstation LAB suite, BradyPrinter i7100
Tiskárny Label Printers
The BradyPrinter i7100 industrial thermal transfer bench top printer is ideal for customers requiring reliable high volume printing on lots of different materials within the aerospace, defence and mass transit, electronics, automotive, logistics, electrical, voice-datacom and lab markets.
  • 300 or 600 dpi
  • Applications include cable and wire identification, circuit board and component identification, and general and industrial identification
  • Mono colour
It guarantees accurate printing on sleeves, tags, labels, tapes, cable markers and much more. TheMore Product Information
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable
Displej Colour touchscreen, 272×480 pixels Colour touchscreen, 272×480 pixels Colour touchscreen, 272×480 pixels Colour touchscreen, 272×480 pixels
Rozhraní Ethernet; I/O connection; Bluetooth (optional); WiFi (optional); Serial; USB Ethernet; I/O connection; Bluetooth (optional); WiFi (optional); Serial; USB Ethernet; I/O connection; Bluetooth (optional); WiFi (optional); Serial; USB Ethernet; I/O connection; Bluetooth (optional); WiFi (optional); Serial; USB
Velikost etikety Height 4 - 6 mm (for the peel-version) Height 4 - 6 mm (for the peel-version) Height 4 - 6 mm (for the peel-version) Height 4 - 6 mm (for the peel-version)
Paměť 257 MB RAM, 50 MB IFFS Flash, SD card, slot up to 512 GB 262 MB RAM, 50 MB IFFS Flash, SD card, slot up to 512 GB 263 MB RAM, 50 MB IFFS Flash, SD card, slot up to 512 GB 256 MB RAM, 50 MB IFFS Flash, SD card, slot up to 512 GB
Napájení AC Adaptor, Lithium-Manganese Battery AC Adaptor, Lithium-Manganese Battery AC Adaptor, Lithium-Manganese Battery AC Adaptor, Lithium-Manganese Battery
Metoda tisku Thermal Transfer Thermal Transfer Thermal Transfer Thermal Transfer
Software Compatible with Brady Workstation, Windows based driver for 3rd party software use Compatible with Brady Workstation, Windows based driver for 3rd party software use Compatible with Brady Workstation, Windows based driver for 3rd party software use Compatible with Brady Workstation, Windows based driver for 3rd party software use
Podporovaný OS Windows® 7, 8, 8.1, 10, Server 2008, 2008 R2, 2012, 2012 R2, 2016 Windows® 7, 8, 8.1, 10, Server 2008, 2008 R2, 2012, 2012 R2, 2016 Windows® 7, 8, 8.1, 10, Server 2008, 2008 R2, 2012, 2012 R2, 2016 Windows® 7, 8, 8.1, 10, Server 2008, 2008 R2, 2012, 2012 R2, 2016
Rozsah teplot 5...40 °C (Operating ) 5...40 °C (Operating ) 5...40 °C (Operating ) 5...40 °C (Operating )