Tlačiť
VWR®, Flat Caps or Plug Caps for VWR® Centrifuge Tubes
Dodávateľ: VWR Collection
525-0923EA 77.31 EUR
525-0923
VWR®, Flat Caps or Plug Caps for VWR® Centrifuge Tubes
Uzávěry Víčka na zkumavky
Purple PP. Sustainable design based on virgin resins.
Closures for use with VWR® 50 ml tubes. Non sterile. Not autoclavable.