Tlačiť
Rack for centrifuge tubes
Dodávateľ: HEATHROW
HS24306
525-0141EA 21.51 EUR
525-0141
Rack for centrifuge tubes
Stojany Tube Racks
ABS
  • Designed for stable use in water baths
  • Supports conical bottom centrifuge tubes
  • Alphanumeric grid reference aids tube identification
  • Sterilisation with 70% alcohol spray
For ABS plastic we do not recommend exceeding 80 °C.
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable